Learn More
Brady™ BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels
BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels are ideal for general purpose barcode applications and withstand liquid nitrogen, autoclave and other harsh environments.
Brand:  Brady™ 148680
 View more versions of this product
Product Code. 17130181
  • 308.00 EUR / Pack of 2000
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
  • Perfect for laboratory identification such as vials, centrifuge tubes and test tubes
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -112 to 158° F (-80 to 70° C)
Compliance
ASTM D 1000;ASTM D 2979;RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
Acrylic
Matte
Thermal Transfer
2000/Pk.
0.75 in.
White
Acrylic
BBP™33 Label Printer
1 in.