Learn More
Brady™ BBP12 Label Printer Kit EU with Unwinder and Workstation PWID Suite
Product Code. 15971032
Click to view available options
:
BBP12 Printer Kit EU PWID; Color Capability: Monochrome; Print Speed: 101.6 mm/s; Print Technology: Thermal Transfer; Resolution: 300 dpi; Weight (Metric): 2.3 kg
Product Code. 15971032
Brand: Brady™ 198595
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This kit contains the BBP12 Label Printer, unwinder, and the Workstation Product & Wire Identification Software suite so that you can immediately start designing professional labels.
The Brady™ BBP™12 gives you the versatility to print all your basic label types from one machine, saving costs, reducing downtime and simplifying your labeling.
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts
  • Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Comes with unwinder and Brady™ Workstation PWID Suite
Specifications
Display Type Color LCD
Print Technology Thermal Transfer
Width (English) 8 in.
Width (Metric) 203 mm
Height (English) 7.5 in.
Height (Metric) 191 mm
Depth (English) 10 in.
Depth (Metric) 259 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes BBP12 Printer, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Brady™ Workstation Product & Wire ID Suite
Languages Czech, Dutch, English, Finnish, French, German, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
Memory 128 MB (Flash), 64 MB (SDRAM), SD Flash Memory Card Up To 32 GB
Model BBP12
Print Speed 101.6 mm/s
Resolution 300 dpi
Connector Types Ethernet, Serial, USB
Operating System Windows™ 7, Windows™ 8, Windows™ 10
Temperature Range (Metric) 5°C to 40°C
Weight (English) 5.1 lb.
Weight (Metric) 2.3 kg
For Use With (Application) Barcode Labeling, Component Labeling, General Identification
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number BBP12-EU-U-PWID