Learn More
Brady™ BBP12 Label printer 300 dpi - Laboratory Kit with BWS Suite
Product Code. 16855741
Click to view available options
Color:
Blue, Grey
Unit Size:
Each
Product Code. 16855741
missing translation for 'mfr': Brady™ 198596
Only null left
Add to basket
Add to basket
The BBP™12 Label Printer packs a lot of punch into one compact tool with its high-speed printing, an extended list of labeling options and a simple, user-friendly design.
  • BBP™12 gives you the versatility to print all your basic label types from one machine - saving costs, reducing downtime and simplifying your labeling
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts, for general identification, wire marking, asset tracking and more
  • Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Ships with a license for the Brady Workstation suite of your preference: Product amd Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
Specifications
Product Type Printer Kit
Color Blue, Grey
Display Type Color LCD
For Use With (Equipment) THT Series Printers, PS Series Printers, 3PS Series Printers, HX Series Printers, 2HX Series Printers
Print Technology Thermal Transfer
Width (English) 7.99 in.
Width (Metric) 202.95 mm
Height (English) 7.5 in.
Height (Metric) 190.5 mm
Depth (English) 10.21 in.
Depth (Metric) 259.33 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monocolour
Display Size 58.42 mm
Includes Unwinder, USB Cable, power cord, product CD, Quick Start Guide,Brady Workstation Software suites, THT-131-461-3, THT-156-492-3, THT-163-499-3, R7950 35mmx70m /O, R6400 65mmx70m / O, R4900 65mmx70m /O
Languages Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
Memory 128 MB Flash, 64 MB SDRAM, SD Flash memory card up to 32 GB
Model BBP12
Print Speed 101.60 mm/sec.
Resolution 300 dpi
Software Type Brady Workstation, Windows based driver for 3rd party software use
Connector Types Ethernet;Serial;USB
Operating System Windows 7;Windows 8;Windows 10
Temperature Range (Metric) 5°C to 40°C
Weight (English) 5.07 lb.
Weight (Metric) 2.3 kg
Certifications/Compliance UL;cUL;CE;RoHS;EAC
For Use With (Application) Labeling
Quantity 1 Ea.
Warranty 1 Year
Power Supply Details AC Adaptor
Voltage 100/240 V
Alternate Part Number BBP12-LAB Kit-EU