missing translation for 'onlineSavingsMsg'
Learn More

BD Ac-DEVD-AMC Caspase-3 Substrate

Product Code. 15848698
Click to view available options
Quantity:
1 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15848698

Brand: BD 556449

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Synthetic tetrapeptide fluorogenic substrate for Caspase-3 (CPP32) containing the amino acid sequence of the PARP cleavage site at Asp-216

Members of the ICE/CED-3 cysteine protease family have key roles in inflammation and mammalian apoptosis. The ICE family member Caspase-3 (also known as CPP32, Yama, apopain) is activated early in apoptosis and appears to be involved in the proteolysis of several important molecules, including poly (ADP ribose) polymerase (PARP). Activated Caspase-3 cleaves PARP from its 116 kDa to an 85 kDa residual fragment. The cleavage site in PARP is C-terminal to Asp-216. The upstream sequence of the cleavage site, DEVD (Asp-Glu-Val-Asp), is utilized as a basis for the highly specific Caspase-3 substrate, Ac (N-acetyl)-DEVD-AMC (7-amino-4-methylcoumarin).

  • Format: Purified
  • Storage buffer: Lyophilized powder

Functional assay

TRUSTED_SUSTAINABILITY

Specifications

Form Lyophilized
For Use With (Application) For Caspase-3, fluorescence quantitation, used in protease assays
Quantity 1 mg
Substrate Type Caspase Substrate
Includes Amino acid sequence of the PARP cleavage site at Asp 216
Molecular Weight (g/mol) 694
Storage Requirements -20°C
Substrate Quantity 1mg

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.