missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Assay Diluent
For reconstituting and diluting standard controls and test samples
Brand: BD 560104
119.02 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Can be used with Flex Set assays
- Storage buffer: aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide

Specifications
Assay Diluent | |
Storage Buffer of aqueous Buffered solution containing fetal bovine Serum and ≤0.09% Sodium Azide | |
30 mL | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Assay Diluent > Size: 30mL
For research use only.
Spot an opportunity for improvement?Share a Content Correction