missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Urea Agar Base
Click to view available options
:
Urea Agar Base; 500g
Specifica
Specifica
| Certifications/Compliance | BAM, CCAM, ISO, USDA |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Urea Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 12mg/L |
| For Use With (Application) | For differentiation of organisms, especially the Enterobacteriaceae, on the basis of Urease production |
| Packaging Type | Bottle |
| Quantity | 500 g |
Sicurezza e movimentazione
Avvertenza
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
- MIXTURE LIST-Contains : urea
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto