Learn More
BD BBL™/Difco™ TB Methylene Blue
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Brand:  BD 212516
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17322441
  • 222.00 EUR / Pack of 4
Estimated Shipment: 13-02-2025
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TB Methylene Blue
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Bottle
4 x 250mL
15° to 30°C
Methylene Blue
For in vitro Diagnostic Use.