Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amoxicillin w/Clavulanic Acid3, AmC-30
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
77.15 EUR
Specifications
Antimicrobial Agents Amoxicillin with Clavulanic Acid
Concentration 20/10 μg
Packaging Type Disc
View More Specs
Products 1
Product Code Brand Antimicrobial Agents Concentration Quantity Price Quantity & Availability  
Product Code Brand Antimicrobial Agents Concentration Quantity Price Quantity & Availability  
11732974
View Documents
BD
231629
Amoxicillin with Clavulanic Acid 20/10 μg 10 Cartridges
77.15 EUR
Pack of 500
Estimated Shipment: 12-02-2025
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Specifications
Amoxicillin with Clavulanic Acid
20/10 μg
Disc
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 548 Days