Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amoxicillin w/Clavulanic Acid3, AmC-30
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
77.15 EUR
Specifications
Antimicrobial Agents | Amoxicillin with Clavulanic Acid |
---|---|
Concentration | 20/10 μg |
Packaging Type | Disc |
Product Code | Brand | Antimicrobial Agents | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Antimicrobial Agents | Concentration | Quantity | Price | Quantity & Availability | |||
11732974
|
BD
231629 |
Amoxicillin with Clavulanic Acid | 20/10 μg | 10 Cartridges |
77.15 EUR
Pack of 500 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Amoxicillin with Clavulanic Acid | |
20/10 μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 548 Days