Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Penicillin (10U)
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
51.15 EUR
Specifications
Antimicrobial Agents | Penicillin |
---|---|
Concentration | 10U |
Packaging Type | Disc |
Description
- Clearly marked on both sides with letters and numbers designating agent and concentration
- Available in pack of 1 and 10
Specifications
Penicillin | |
10U | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 730 Days