Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Sulfamethoxazole-Trimethoprim
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
52.30 EUR
Specifications
Antimicrobial Agents Sulfamethoxazole, Trimethoprim
Packaging Type Disc
View More Specs
Products 1
Product Code Brand Concentration Quantity Price Quantity & Availability  
Product Code Brand Concentration Quantity Price Quantity & Availability  
11757594
View Documents
BD
231539
23.75μg/1.25μg
10/Pk.
52.30 EUR
Pack of 10
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
Description
Description
  • Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Sulfamethoxazole, Trimethoprim
Disc
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 1095 Days