missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BD Biosciences Fixable Viability Stain 620
For discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications
Brand: BD 564996
244.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reacts with and covalently binds to cell-surface and intracellular amines
- Permeable plasma cell membranes, such as those present in necrotic cells, allow intracellular diffusion of dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells
- Necrotic cells present in a typical in vitro assay label with higher levels of dye increases their fluorescence intensity 10-20 fold over viable cells
- Labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeabilization and subsequent intracellular staining while maintaining stable viability stain fluorescence
- Excited by Yellow-Green laser (with an excitation maximum of 523nm), but is also well-excited by the Blue laser
- Fluorescence emission maximum of 617nm
- Applications: Flow cytometry, Intracellular staining
Specifications
100 μg | |
Flow cytometry, Intracellular staining (flow cytometry) (Tested During Development) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction