missing translation for 'onlineSavingsMsg'
Learn More

BD C5a, Recombinant mouse

Bioactive cleavage recombinant protein released from plasma component C5 during complement activation

Brand:  BD 622597

322.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15886279

  • 323.00 EUR / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Anaphylatoxin and potent pro-inflammatory agent
  • Responsible for mediation of variety of cellular immune responses
  • Purified by affinity chromatography
  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA and ProClin 150

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Purified Recombinant Mouse C5a
10 Tests
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For research use only.