Learn More
BD Cytometric Bead Array (CBA) Human IL-1β Enhanced Sensitivity Flex Set (Bead B4)
Lyophilized recombinant Human IL-1β measures soluble analytes present in very low concentrations
Brand:  BD 561509
Product Code. 15831089
  • 419.00 EUR / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • Capture bead
  • Detection reagent
  • Standard
Description
Description
  • Set is a bead-based immunoassay capable of measuring human interleukin-1β (IL-1β) in serum, plasma and cell culture supernatant samples
  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Includes 100 tests
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-1beta
1 Vial capture bead, 1 vial detection reagent and 2 vials standard
Serum, Plasma, Cell Culture Supernatant
Measures Human interleukin-1beta
Human
100 Tests
4°C
Human IL-1beta enhanced sensitivity flex set (bead B4)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • BD Cytometric Bead Array (CBA) Human IL-1
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT