Learn More
BD Cytometric Bead Array (CBA) Mouse CD62L (L-Selectin); Flex Set, Bead E7
Bead-based immunoassay capable of measuring mouse soluble CD62L (L-Selectin) in cell culture supernatant samples
Brand: BD 560385
1070.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- Mouse soluble CD62L (L-Selectin) Standard
- Mouse soluble CD62L (L-Selectin) PE Detection Reagent
- Mouse soluble CD62L (L-Selectin) Capture Bead E7
Description
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution.
- Mouse reactivity was determined by testing samples with this Felx set
- Must be used in conjunction with CBA Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifications
CD62L | |
Mouse soluble CD62L (L-Selectin) standard, Mouse soluble CD62L (L-Selectin) PE detection reagent, Mouse soluble CD62L (L-Selectin) capture bead E7 | |
4°C | |
Mouse CD62L (L-Selectin) flex set (bead E7) |
Mouse | |
100 Tests | |
Flow Cytometry |