missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-12/IL-23P40; Flex Set, Bead D7

Product Code. 15849968
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15849968

Brand: BD 560151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Mouse IL-12/IL-23P40, Standard (two vials), Mouse IL-12/IL-23P40 PE Detection Reagent (one vial), Mouse IL-12/IL-23P40; Capture Bead D7 (one vial)

Bead-based immunoassay for measuring mouse p40 subunit of interleukin-12/Interluekin-23 (IL-12/IL-23p40) in serum and cell culture supernatant samples

  • Mouse reactivity was determined by testing samples with Flex set
  • Theoretical limit of detection is 13.6pg/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12/IL-23P40
Cross Reactivity Mouse
Includes Mouse IL-12/IL-23P40, standard (2 vials), Mouse IL-12/IL-23P40 PE detection reagent (one vial), Mouse IL-12/IL-23P40, capture bead D7 (one vial)
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IL-12/IL-23p40 flex set (bead D7)

Research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.