Learn More
BD Cytometric Bead Array Mouse IL-6 Enhanced Sensitivity Flex Set
For measuring mouse Interleukin-6 (IL-6) in serum and cell culture supernatant samples
Brand: BD 562236
Includes: Capture beads, detection reagent, standards.
Description
- Bead-based immunoassays
- Bead B4
- Also measures soluble analytes present in very low concentrations
Specifications
IL-6 | |
1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) | |
Serum, Cell Culture Supernatant | |
Measures Mouse interleukin-6, flow cytometry |
Mouse | |
100 Tests | |
4°C | |
Mouse IL-6 enhanced sensitivity flex set (bead B4) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BD Cytometric Bead Array (CBA) Mouse IL-6 Enhanced Sensitivity Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT