missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Antibiotic Medium 19
For determining antibiotic potency by the microbiological assay technique. BD Difco™ Dehydrated Culture Media: Antibiotic Medium 19 is part of a large selection of dehydrated media.
Brand: BD 243100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Dehydrated
- Culture Media
- Antibiotic Medium 19
Spezifikation
EP, USP | |
Dehydrated | |
Antibiotic Medium | |
Microbiological assay of Antibiotics | |
500 g |
<30°C | |
Antibiotic Medium 19 | |
Beef extract 2.4g/L, Yeast extract 4.7g/L, Peptone 9.4g/L, Dextrose 10g/L, Sodium Chloride 10g/L, Agar 23.5g/L | |
Bottle |