missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MacConkey Broth

Product Code. 11748303
Click to view available options
:
MacConkey Broth; 500g
This item is not returnable. View return policy

Product Code. 11748303

Brand: Fisherbrand™ 220100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A selective medium for detecting gram-negative, lactose-fermenting coliforms. Fisherbrand™ MacConkey Broth is provided dehydrated.

  • 500g bottle
  • Works excellently with milk or water

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, EP, ISO, JP, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Broth
Product Type Microbiology Media
Formulation Oxgal 5g/L, Pancreatic digest of Gelatin 20g/L, Lactose 10g/L, Bromcresol Purple 0.01g/L
For Use With (Application) Detection of Coliform organsms in Milk and Water
Packaging Type Bottle
Quantity 500 g
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : oxgall
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.