Learn More
BD Difco™ Dehydrated Culture Media: Tomato Juice Agar
Click to view available options
:
Tomato Juice Agar; 500g
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Tomato Juice Agar |
| Product Type | Agar |
| Formulation | Tomato Juice (from 400mL) 20g/L, Peptone 10g/L, Peptonized Milk 10g/L, Agar 11g/L |
| For Use With (Application) | Isolation of Lactobacillus species |
| Packaging Type | Bottle |
| Quantity | 500 g |