missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD ELISA Set
Click to view available options
Sufficient For:
5 Plates of 96-well
Unit Size:
Each
Description
Includes
- Capture Antibody: Purified anti-human C5b-9 (250X)
- Detection Antibody: Biotin anti-human C5b-9 (250X)
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X)
- Standards: Human C5b-9 Standard (5 vials)
- Allows flexible assay design to fit individual laboratory needs
- Immunoassay is calibrated against purified Cobra Venom-Factor activated human serum
- Immunoassay with different sensitivity and dynamic range
ELISA
Specifications
Specifications
| Product Type | Human C5b-9 ELISA set |
| Sufficient For | 5 Plates of 96-well |
| For Use With (Application) | To develop ELISA for natural C5b-9 in Serum, Plasma and other biological samples |
| Kit Contents | Biotin anti-Human C5b-9 (250X) (detection) Antibody, Human C5b-9 Standard (5 vials) (standard), Instruction/analysis certificate, Purified anti-Human C5b-9 (250X) (capture) Antibody, Streptavidin-HRP Conjugate (250X) |
| Quantity | 5 Plates |
| Storage Requirements | 4°C |
Safety and Handling
Product Identifier
- Human C5b-9 ELISA Set
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : cmit/mit
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction