missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD279 (PD-1)
CD279 is an immunoregulatory receptor expressed on activated T cells, B cells, and myeloid cells. BD Horizon™ BV480 Mouse Anti-Human CD279 (PD-1) contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms.
Brand: BD 566112
This item is not returnable.
View return policy
Description
With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter setBV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD279 (PD-1) | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
100 Tests | |
QC Testing: Human | |
5 μL | |
5133 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction