missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human CD253 (TRAIL) ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
Each
Description
Includes
- Capture Antibody: Anti-human CD253 monoclonal antibody
- Detection Antibody: Biotinylated anti-human CD253 monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant human TRAIL, lyophilized
- Instruction / Analysis Certificate: (lot-specific)
Requires
96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, microplate reader capable of measuring absorbance at 450nm, precision pipettes, graduated cylinder, one liter, deionized or distilled water, wash bottle or automated washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, plate sealers or parafilm
- Alias: TRAIL
- For natural or recombinant human TRAIL in serum, plasma, and cell culture supernatants
Enzyme Linked Immunosorbent Assay
Specifications
Specifications
| Product Type | Human CD253 (TRAIL) ELISA set |
| Sufficient For | 20 x 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human TRAIL in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human CD253 monoclonal (capture) Antibody, Biotinylated anti-Human CD253 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human TRAIL (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction