Learn More
BD Human IL-8 ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand:  BD 555244
1207.67 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15868028
  • 1210.00 EUR / 1 set
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/ analysis certificate
Description
Description
To develop enzymelinked immunosorbent assays (ELISA) for natural or recombinant human IL-8 in serum, plasma, and cell culture
supernatants
Enzyme-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3.1 to 200pg/mL
20 96-well Plates
anti-Human IL-8 monoclonal (capture) Antibody, Biotinylated anti-Human IL-8 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-8 (standard), Streptavidin-HRP Conjugate
2°C to 8°C
Human IL-8 ELISA set
To develop ELISA for natural or recombinant Human IL-8 in Serum, Plasma and cell culture supernatants
20 Plates