Learn More
BD Human Mesenchymal Stem Cell Analysis Kit
For reproducible multicolor phenotyping of MSC expansions
Brand: BD 562245
2215.79 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- PE hMSC Negative Cocktail
- PE hMSC Isotype Control Negative Cocktail
- hMSC Positive Cocktail
- hMSC Isotype Control Positive Cocktail
- FITC Mouse Anti-Human CD90
- PE Mouse Anti-Human CD44
- PE Mouse IgG2b, k Isotype Contro
- APC Mouse Anti-Human CD73
- PerCP-Cy5.5 Mouse Anti-Human CD105 (Endoglin)
Description
Provides comprehensive system for the reliable, in-depth characterization of human bone marrow-derived multipotent mesenchymal stromal cells (BM-MSCs) based on the phenotypic signature described by the The Mesenchymal and Tissue Stem Cell Committee of the International Society for Cellular Therapy (ISCT).
- Modular designed for additional markers to be “dropped in”
- Contains pre-conjugated and pre-titrated cocktails of the ISCT-defined positive expression markers (CD105 PerCP-Cy5.5/CD73 APC/CD90 FITC) and negative expression marker
- Enables the easy addition of supplementary monoclonal antibodies against critical cell surface marker
Specifications
50 Tests | |
4°C | |
For reproducible multicolor phenotyping of MSC expansions, FCM | |
Human |