missing translation for 'onlineSavingsMsg'
Learn More

BD HumanIFN-γ ELISA Set

Product Code. 15868008
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15868008

Brand: BD 555142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Capture Antibody: Anti-mouse IFN-Γ monoclonal antibody
  • Detection Antibody: Biotinylated anti-mouse IFN-Γ monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant mouse IFN-Γ, lyophilized
  • Instruction / Analysis Certificate

For immunoassay protocols

The OptEIA set for human interferon-gamma (IFN-Γ), contains components necessary to develop enzyme-linked immunosorbent assays (ELISA)

  • Reactivity: Human
  • Lyophilized
  • For natural or recombinant human IFN-Γ in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4.7 to 300pg/mL
Product Type Human IFN-ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Human IFN-g in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse IFN-G monoclonal (capture) antibody, Biotinylated anti-Mouse IFN-G monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IFN-G (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.