Learn More
BD Monolight™ Luminometer Cuvettes
For multi-application gel imaging
Brand: BD 556862
98.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Plastic
- For use with BD™ Monolight™ Luminometers
- Size: 12 x 75mm
Specifications
Luminometer Cuvette | |
Each | |
For multi-application gel imaging |
Plastic | |
For luminometer |