missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse GM-CSF ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
The OptEIA™ Set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse GM-CSF in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-mouse GM-CSF
- Detection antibody: biotinylated anti-mouse GM-CSF
- Enzyme reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standard: lyophilized recombinant mouse GM-CSF
- Standard range: 15.6 - 1000 pg/mL
ELISA
Specifications
Specifications
| Concentration | 15.6 to 1000pg/mL |
| Product Type | Mouse GM-CSF ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse GM-CSF in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Mouse GM-CSF monoclonal (capture) antibody, Biotinylated anti-Mouse GM-CSF monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse GM-CSF (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction