missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse IgE ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
Includes
Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
To develop enzyme-linked immunosorbent assays (ELISA) for mouse IgE in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Concentration | 1.6 to 100ng/mL |
| Product Type | Mouse IgE ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for Mouse IgE in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Mouse IgE monoclonal (capture) antibody, Biotinylated anti-Mouse IgE monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized purified Mouse IgE (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction