Learn More
BD Mouse IL-12 (p70) ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand: BD 555256
949.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-12 (p70) in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
Specifications
62.5 to 4000pg/mL | |
20 96-well Plates | |
20 Plates |
Mouse IL-12 (p70) ELISA set | |
To develop ELISA for natural or recombinant Mouse IL-12 (p70) in Serum, Plasma and cell culture supernatants | |
2°C to 8°C |