missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse interleukin-2 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-2 in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-mouse IL-2 monoclonal antibody
- Detection antibody: biotinylated anti-mouse IL-2 monoclonal antibody
- Enzyme reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: lyophilized recombinant mouse IL-2
- Standard range: 3.1 - 200 pg/mL
ELISA
Specifications
Specifications
| Concentration | 3.1 to 200pg/mL |
| Product Type | Mouse IL-2 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse IL-2 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Mouse IL-2 monoclonal (capture) antibody, Avidin-HRP, Biotinylated anti-Mouse IL-2 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-2 (standard) |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Safety and Handling
Product Identifier
- Mouse IL-2 OptEIA Set
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : CMIT/MIT
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction