Learn More
BD Mouse TNF (Mono/Mono) ELISA Set
Product Code. 15818038
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Toxic if ingested, inhaled, or in contact with skin
Product Code. 15818038
Brand: BD 555268
Toxic if ingested, inhaled, or in contact with skin
Includes
Capture Antibody: Anti-mouse TNF-α , Detection Antibody: Biotinylated anti-mouse TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate, Standard: Recombinant mouse TNF-α (lyophilized)
Requires
96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, microplate reader capable of measuring absorbance at 450nm, precision pipettes, graduated cylinder, deionized or distilled water, wash bottle or automated washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, plate sealers or parafilm
Determination of mouse tumor necrosis factor
  • For natural or recombinant mouse TNF in serum, plasma, and cell culture supernatants
  • Allow flexible assay design
  • Calibrated against purified Baculovirus-expressed recombinant mouse TNF
  • Intended for use as integral unit
ELISA
TRUSTED_SUSTAINABILITY
Specifications
Concentration 615.6 to 1000pg/mL
Product Type Mouse TNF (Mono/Mono) ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse TNF in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse TNF monoclonal (capture) antibody, Biotinylated anti-Mouse TNF monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse TNF (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C