Learn More
BD Native Protein
Useful as a standard in a sandwich ELISA to measure mouse C3a protein levels
Brand: BD 558618
Description
Purified Mouse C3a protein is a native protein isolated from activated mouse serum. Anaphylatoxin C3a is a bioactive cleavage product released from plasma component C3 during complement activation and is involved in mediation of a variety of cellular immune responses, as well as being a potent pro-inflammatory agent. The release of this cleavage product is a reliable indicator of in vivo or in vitro complement activation.
- Storage buffer: Lyophilized in an aqueous buffered solution containing BSA and ProClin 150
- Isolated and purified by affinity chromatography
ELISA
Specifications
ELISA | |
Mouse C3a Native Protein | |
In vivo or in vitro complement | |
10 Tests | |
4°C | |
Recombinant |
Lyophilized in an aqueous Buffered solution containing BSA and ProClin 150 | |
6.5 | |
Affinity Chromatography | |
Mouse Serum | |
Mouse | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research use only.