Learn More
BD Phosflow™ Violet Fluorescent Cell Barcoding Kit
Provides reagents for successfully barcoding T cell subsets from human whole blood samples for studying phosphorylated protein induction in MAP Kinase and JAK/STAT signaling pathways
Brand:  BD 561570
1599.14 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15851139
  • 1600.00 EUR / Each
Explore more special offers
Includes: Cell Barcoding Dye 450 (CBD450) and Cell Barcoding Dye 500 (CBD500), Fluorescent Cell Barcoding Wash Buffer, and choice of one Alexa Fluor™ 647-conjugated, phospho-specific antibody
Description
Description
  • Reactivity: human
  • Size: 800 tests
TRUSTED_SUSTAINABILITY
Specifications
Specifications
800 Tests