missing translation for 'onlineSavingsMsg'
Learn More

BD Rat MCP-1 ELISA Set

Product Code. 15838008
Click to view available options
Quantity:
20 Plates
missing translation for 'unitSize'
1 set
This item is not returnable. View return policy

Product Code. 15838008

missing translation for 'mfr': BD 555130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For rat monocyte chemoattractant protein-1

The OptEIA™ set contains components necessary to develop enzymelinked immunosorbent assays (ELISA) for natural or recombinant rat MCP-1 in serum, plasma, and cell culture supernatants.

  • Capture antibody: anti-rat MCP-1 monoclonal antibody
  • Detection antibody: biotinylated anti-rat MCP-1 monoclonal antibody
  • Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
  • Standards: lyophilized recombinant rat MCP-1
  • Standard range: 31.3 - 2000pg/mL

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 31.3 to 2000pg/mL
Product Type Rat MCP-1 ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Rat MCP-1 in Serum, Plasma and cell culture supernatants
Kit Contents anti-Rat MCP-1 (capture) antibody, Biotinylated anti-Rat MCP-1 (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Rat MCP-1 (standard), Streptavidin-HRP
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.