missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Rat Monocyte chemoattractant protein-1 (MCP-1)
Highly glycosylated protein with chemoattractant activity for monocytes, lymphocytes and basophils
Brand: BD 555110
438.28 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Member of C-C chemokine superfamily
- Homolog of human MCP-1
- Reactivity: rat
- Size: 27 - 30kD
- Expressed by variety of cell types including monocyte/macrophages, endothelial cells and mesangial cells
- Supplied as frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution
- Contains glycerol, 1.0mg/mL biotechnology grade, low endotoxin bovine serum albumin
- Storage buffer: Frozen aqueous buffered solution containing BSA and glycerol
Specifications
0.1 mg/mL | |
ELISA standard, bioassay, intracellular block/flow cytometry | |
5 μg |
PharMingen™ Recombinant Rat MCP-1 | |
Glycerol 1.0mg/mL biotechnology grade, low endotoxin bovine Serum Albumin | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction