Learn More
Bel-Art™ Glove Box Economy Sleeved Gloves Product Code.: 12398617
Add to basket
Bel-Art™ Glove Box Economy Sleeved Gloves
Product Code. 12398617 Shop All SP Scienceware Products
Large (size 10)
Click to view available options
Large (size 10)
Product Code. 12398617
Brand: Bel-Art™ H500290000
Add to basket
Add to basket
Compatible with Scienceware Portable Glove Box or any glove box with 6 in. glove ports
  • Durable neoprene fabrication
  • Safeguards against gas, liquid, or solid substance penetrants
  • Use with Clamping Ring, Part No. 03-421-102
  • Large, size 10
  • 1 pair/Pk.
Specifications
Color Black
Hand Specific
Latex Free No
Material Bonded Neoprene
Product Type Glove Box Gloves
Size 10
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Bel-Art™ Glove Box Economy Sleeved Gloves >