Learn More
Bel Art™ Secador™ Standard Desiccator Cabinet, Vertical Model
Shop All SP Scienceware Products
Click to view available options
Height (Metric):
31.4 cm
41.7 cm
51.8 cm
Color:
Amber
Blue with Clear Door
Clear
Clear with Blue Ends
No. of Shelves:
2
3
Volume (English):
1.17 cu. ft.
1.58 cu. ft.
1.9 cu. ft.
Unit Size:
Each
Description
Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Compact 34.1 W x 41.4cm D footprint for space saving stacking up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
- Can be used with desiccant cartridges, F42049-0100 or F42046-0000
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. F42049-0100, F42046-0000
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Cabinet Style | Vertical |
| Material | Copolyester |
| Product Type | Desiccator |
| Door Style | Left-hinged and gasketed |
| For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant |
| Includes | Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer |
| Model | 2.0 |
| No. of Shelves | 2 |
| Show More |