Learn More
Bel-Art™ SP Scienceware™ Slide Boxes
Will not warp, splinter or crack, and are unaffected by moisture. Bel-Art™ SP Scienceware™ Slide Boxes are compact, durable slide boxes to provide the maximum protection for 25 x 75mm (1 x 3 in.) microscope slides.
47.40 EUR
Specifications
For Use With (Equipment) Microscope Slide
Includes Numbered index on base
View More Specs
Products 1
Product Code Brand Slide Capacity Price Quantity & Availability  
Product Code Brand Slide Capacity Price Quantity & Availability  
15515614
View Documents
Bel-Art™
H44577-0000
100
47.40 EUR
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
Description
Description
  • Heavy-walled boxes are rugged and will not bend, split or chip
  • Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
  • Easily identify slides with numbered index included on base and cover
Specifications
Specifications
Microscope Slide
Numbered index on base