Learn More
Bel-Art™ SP Scienceware™ Y Connectors
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
Brand:  Bel-Art™ F19614-0000
 View more versions of this product
Product Code. 12373720
  • 56.15 EUR / Pack of 12
Estimated Shipment: 28-02-2025
Description
  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
Autoclavable
6mm
Y Tubing Connector
Polypropylene
Y