Learn More
Kern Sohn Bench Scale FCE
Entry-level bench scale that is mobile, practical, and lightweight.
Brand:  Kern Sohn FCE 3K1N
Encompass_Preferred
 View more versions of this product
Product Code. 17361633
  • 137.70 EUR / Each
Description
  • Simple and convenient 2-key operation
  • Very fast display: steady weight values within 3 s
  • Ideal as a compact letter and parcel scale, especially where space is limited
  • Ideal as a grading, commissioning or simple check-weighing scale in production or dispatch
  • Protective working cover included with delivery
Specifications
Specifications
3 kg
228 x 252 mm
2 g