missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Benchtop pH/mV Meter with 0.01 Resolution

Product Code. 11682220
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11682220

Brand: Hanna Instruments™ HI2211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Features automatic calibration to 1 or 2 points with 5 preprogrammed buffers. Hanna Instruments™ Benchtop pH/mV Meter with 0.01 Resolution displays pH and mV readings simultaneously and comes complete with an electrode, probe, buffers, electrolyte, cleaning solution and probe holder.

With the HI2211, operation is simplified to calibration, mode selection (pH or mV), and a Last Stored Reading feature. The mV mode is used with an optional ORP electrode that is used to check the oxidation or reduction potentials of a solution. The mV mode of the HI2211 is also useful in checking the offset and slope of the pH electrode, which is helpful in knowing if the probe is functioning correctly.

  • Easy to use with simple buttons and a large, clear display with stability indicator
  • Temperature Compensation: Automatic or manual from -9.9 to 120°C
  • Measurement Recall: Press the MEM button to store a reading, and recall it anytime by pressing MR
  • Separate inputs for a universal BNC pH electrode and thermistor temperature probe
  • High accuracy (0.5°C) temperature probe for precise temperature compensation
  • Automatic calibration to 1 or 2 points using the 5 preprogrammed buffers (4.01, 6.86, 7.01, 9.18, and 10.01)
  • mV mode can be used with a variety of ORP electrodes
  • HI1131B High Temperature (HT) glass body, double junction, refillable pH electrode with indicating sensor
  • Specifications

    Includes HI1131B pH electrode, HI7662 temperature probe, HI76404N electrode holder, HI70004 pH 4.01 buffer solution sachet, HI70007 pH 7.01 buffer solution sachet, HI7082 electrolyte solution, HI700601 cleaning solution sachet, 12 VDC adapter, and instructions
    Accuracy (Temperature) ±0.5°C/±1°F
    Accuracy (mV) ±0.2 (±399.9), ±1 (±2000)
    Accuracy (pH) ±0.01
    Calibration Automatic
    Range (pH) -2 to 16
    Resolution (pH) 0.01
    Range (mV) ±399.9, ±2000
    Calibration Points 1 or 2
    Temperature Range (Metric) -9.9°C to +120°C
    Temperature Range (English) 14.2°F to 248°F
    Display Type LCD
    Dimensions (L x W x H) 240 x 182 x 74 mm (9.4 x 7.1 x 2.9 in.)
    Warranty 2 Years (6 Months Probe)
    Material High Temperature glass (electrode), Stainless Steel (temperature probe)
    Power Supply 12 VDC adapter
    Probe Type HI1131B pH electrode (glass body, double junction, refillable, with an indicating sensor), HI7662 temperature probe (stainless steel)
    Relative Humidity Range Max. 95%
    Temperature Compensation Automatic or manual, -9.9 to 120°C
    Temperature (English) Operating 32°F to 122°F
    Temperature (Metric) Operating 0°C to 50°C
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.