Learn More
Benzoic anhydride, 98%, Thermo Scientific Chemicals
22.40 EUR - 385.00 EUR
Chemical Identifiers
CAS | 93-97-0 |
---|---|
Molecular Formula | C14H10O3 |
Molecular Weight (g/mol) | 226.23 |
MDL Number | MFCD00003073 |
InChI Key | CHIHQLCVLOXUJW-UHFFFAOYSA-N |
Synonym | benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate |
PubChem CID | 7167 |
ChEBI | CHEBI:38815 |
IUPAC Name | benzoyl benzoate |
SMILES | C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697432
|
Thermo Scientific Acros
|
25 g | Plastic bottle |
22.40 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297493
|
Thermo Scientific Acros
105501000 |
100 g | Plastic bottle |
38.20 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10204660
|
Thermo Scientific Acros
|
500 g | Plastic bottle |
117.00 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994971
|
Thermo Scientific Acros
|
2.5 kg | Plastic bottle |
385.00 EUR
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-97-0 | |
226.23 | |
CHIHQLCVLOXUJW-UHFFFAOYSA-N | |
7167 | |
benzoyl benzoate |
C14H10O3 | |
MFCD00003073 | |
benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate | |
CHEBI:38815 | |
C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 |
Specifications
93-97-0 | |
100.0 | |
White | |
360.0°C | |
Authentic | |
C14H10O3 | |
MFCD00003073 | |
01,49; 05,23 | |
1.19 | |
Solubility in water: 0,01g/L. Other solubilities: soluble in alcohol,chloroform,acetone,benzene,,ethyl acetate,toluene,xylene,ether,glacial,acetic acid,acetic anhydride,moderately soluble in petroleum ether | |
C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 | |
226.23 | |
CHEBI:38815 | |
98% | |
Benzoic anhydride, 98% |
97.5 | |
38.0°C to 44.0°C | |
1.1900g/mL | |
>112°C | |
97.5% min. (HPLC) | |
(C6H5CO)2O | |
09,164 | |
15,1094 | |
benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate | |
CHIHQLCVLOXUJW-UHFFFAOYSA-N | |
benzoyl benzoate | |
7167 | |
226.23 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-291-1
missing translation for 'tsca' : TSCA
- Benzoic anhydride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only