Learn More
Benzothiazole-2-carbonyl chloride, 95%, Thermo Scientific Chemicals
59.30 EUR - 151.76 EUR
Chemical Identifiers
CAS | 67748-61-2 |
---|---|
Molecular Formula | C8H4ClNOS |
Molecular Weight (g/mol) | 197.636 |
MDL Number | MFCD03659697 |
InChI Key | AOIGQLLPWDXVGB-UHFFFAOYSA-N |
Synonym | benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride |
PubChem CID | 2776254 |
IUPAC Name | 1,3-benzothiazole-2-carbonyl chloride |
SMILES | C1=CC=C2C(=C1)N=C(S2)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305425
|
Thermo Scientific Alfa Aesar
L19631.MD |
250 mg |
59.30 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395415
|
Thermo Scientific Alfa Aesar
L19631.03 |
1 g |
151.76 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67748-61-2 | |
197.636 | |
AOIGQLLPWDXVGB-UHFFFAOYSA-N | |
2776254 | |
C1=CC=C2C(=C1)N=C(S2)C(=O)Cl |
C8H4ClNOS | |
MFCD03659697 | |
benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride | |
1,3-benzothiazole-2-carbonyl chloride |
Specifications
67748-61-2 | |
C8H4ClNOS | |
UN3261 | |
Moisture sensitive | |
AOIGQLLPWDXVGB-UHFFFAOYSA-N | |
1,3-benzothiazole-2-carbonyl chloride | |
2776254 | |
95% |
113°C to 115°C | |
MFCD03659697 | |
130990 | |
benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride | |
C1=CC=C2C(=C1)N=C(S2)C(=O)Cl | |
197.636 | |
197.64 | |
Benzothiazole-2-carbonyl chloride |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.