missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Tubulin Rabbit anti-Human, Mouse, Rat, Zebrafish, CoraLite 555, Clone: 2O13, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL555-80713-100UL
This item is not returnable.
View return policy
Description
Beta Tubulin antibody apecifically detetects Beta Tubulin antigen in Human, Mouse, Rat, Zebrafish Samples. It is validated for Immunofluorescence.Specifications
Beta Tubulin | |
2O13 | |
CoraLite 555 | |
BC000748 | |
Rabbit | |
Protein A purification | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Liquid |
Immunofluorescence | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
TUBB3 | |
Beta Tubulin fusion protein Ag0117 | |
100 μL | |
10381 | |
Store at -20°C. Avoid exposure to light. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction