missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betulin, 95.2%, MP Biomedicals™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 473-98-3 |
Color | White |
Assay Percent Range | 95.2% |
Molecular Formula | C30H50O2 |
Quantity | 1 g |
Synonym | betulin, betulinol, betuline, trochol, betulol, lup-20 29-ene-3b,28-diol, lup-20 29-ene-3beta,28-diol, unii-6w70hn7x7o, lup-20 30-ene-3beta,28-diol, lup-20 29-ene-3,28-diol, 3beta |
InChI Key | FVWJYYTZTCVBKE-ROUWMTJPSA-N |
SMILES | CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)CO |
IUPAC Name | (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a-(hydroxymethyl)-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysen-9-ol |
Molecular Weight (g/mol) | 442.728 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'