missing translation for 'onlineSavingsMsg'
Learn More

Bevirimat, MedChemExpress

Product Code. p-200095942
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16491528

Brand: Medchem Express HYN0842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bevirimat (PA-457; MPC-4326; YK FH312) is an anti-HIV drug derived from a betulinic acid-like compound; is believed to inhibit HIV by a novel mechanism, so-called maturation inhibition.

Bevirimat (PA-457

Specifications

Chemical Name or Material Bevirimat
Physical Form Solid
CAS 174022-42-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C36H56O6
For Use With (Application) COVID-19-anti-virus
Synonym PA-457 MPC-4326 YK FH312
Solubility Information DMSO : ≥ 50 mg/mL (85.49 mM)
SMILES OC([C@]1(CC[C@H]2C(C)=C)[C@@]2([H])[C@](CC[C@@]3([H])[C@]4(CC[C@]5([H])[C@@]3(CC[C@H](OC(CC(C)(C)C(O)=O)=O)C5(C)C)C)C)([H])[C@@]4(C)CC1)=O
Molecular Weight (g/mol) 584.83
Formula Weight 584.83
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.