Learn More
Thermo Scientific™ Bile Esculin Azide Agar
Isolate, enumerate and presumptively identify group D streptococci and enterococci with a medium that utilizes bile salts to improve selectivity.
12.55 EUR
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
10382223
|
Thermo Scientific™
PO5062A |
Bile Esculin Azide Agar | 90mm Monoplate | 10/Pk. |
12.55 EUR
Pack of 10 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||
Description
Isolate, enumerate and presumptively identify group D streptococci and enterococci using Thermo Scientific™ Bile Esculin Azide Agar, a microbiological media that includes a high portion of Ox bile.
Ox bile present in the media inhibits the growth of Gram-positive bacteria except Enterococci, while Sodium azide suppresses the Gram-negative bacteria. Group D streptococci and enterococci are able to hydrolyse aesculin into aesculetin and glucose, and the bondage of aesculetin and ferric ammonium citrate causes the brown-black to black halo around the colonies.Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Bile Esculin Azide Agar | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 14 Weeks