Learn More
Thermo Scientific™ BioBasic™ 18 HPLC Columns
Product Code. p-6084361 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
20 mm
Unit Size:
Each
Pack of 4
Product Code. 10434773
Brand: Thermo Scientific™ 72105254630
Only null left
Add to basket
Add to basket
Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.
Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY
Specifications
Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Analytical Column
Technique Bio-LC