Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
579.94 EUR - 2663.09 EUR
Specifications
Pore Size | 3 μm |
---|---|
Type | Membrane |
For Use With (Application) | Endothelial Cell Invasion |
No. of Wells | 24 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11543540
|
Corning™
354142 |
5/Pk. |
2663.09 EUR
Pack of 5 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
10290862
|
Corning™
354141 |
1/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
3 μm | |
Membrane | |
Endothelial Cell Invasion | |
24 |