Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
Enables high strength biomagnetic separation
1870.00 EUR
Specifications
For Use With (Equipment) | 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer |
---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11997411
|
Agilent BioTek
7102216 |
96-Well Ring Magnet |
1870.00 EUR
Each |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Magnet ring
- External
- For insertion into washer biomagnetic separation carriers
- Beads pulled to 4-zone ring at well bottom
Specifications
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer |